Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rubidium nitrate 99.975% (metals basis), Puratronic®

Supplier: Origen Biomedical, Inc
Danger

010852-18 010852-09
AA10852-18EA 714 USD InStock
AA10852-18 AA10852-09
Rubidium nitrate 99.975% (metals basis), Puratronic®
Rubidium nitrate
MDL: MFCD00011193

Purity: 99.975% CAS No.: 13126-12-0 Molecular Formula: RbNO3 Formula Weight: 147.47 Physical Form: Crystalline Density: 3.11 Refractive Index: 1.52 Sensitivity: Hygroscopic Melting Point: 310° UN No.: 1477 MDL No.: MFCD00011193


Formula: NO₃Rb
Boiling Pt: 578 °C (1013 hPa)
Melting Pt: 310 °C
Density: 3.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011193
CAS Number: 13126-12-0
EINECS: 236-060-1
UN: 1477
ADR: 5.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty