011827-30
011827-18
AA11827-30EA
410.83
USD
InStock
AA11827-30
AA11827-18
Phosphotungstic acid hydrate, REacton®, Reagent Grade
Phosphotungstic acid hydrate
MDL: MFCD00011342
CAS No.: 12501-23-4
Molecular Formula: H3PO4.12WO3.xH2O
Formula Weight: 2880.20(anhy)
Physical Form: Powder
Sensitivity: Moisture and Light Sensitive
Appearance: Off-white to pale yellow
Melting Point: 95°
UN No.: 3260
MDL No.: MFCD00011342