Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc ≥99.994% (metals basis), wire, Puratronic®, Diameter 0.25 mm (0.01 in)

Supplier: Origen Biomedical, Inc

012055-G6 012055-G2 012055-H2
AA12055-G6EA 237 USD InStock
AA12055-G6 AA12055-G2 AA12055-H2
Zinc ≥99.994% (metals basis), wire, Puratronic®, Diameter 0.25 mm (0.01 in)
Zinc
MDL: MFCD00011291

Purity: 99.994% CAS No.: 7440-66-6 Physical Form: ≈0.35g/m Odor: Odorless MDL No.: MFCD00011291


Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty