Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium hexahydroxoplatinate ≥99.9% (metals basis)

Supplier: Origen Biomedical, Inc
Warning

012620-03 012620-06
AA12620-03EA 333 USD InStock
AA12620-03 AA12620-06
Disodium hexahydroxoplatinate ≥99.9% (metals basis)
Disodium hexahydroxoplatinate
MDL: MFCD00014236 Notes: Pt 57%

Purity: 99.9% CAS No.: 12325-31-4 Molecular Formula: H6Na2O6Pt Formula Weight: 343.10 Physical Form: Crystalline Appearance: Yellow to brown Melting Point: 150° dec. MDL No.: MFCD00014236


Formula: H₆Na₂O₆Pt
MW: 343.1
Melting Pt: 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014236
CAS Number: 12325-31-4
EINECS: 235-590-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty