Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hexaiodoplatinate(IV) Pt ≥ 18,2%

Supplier: Origen Biomedical, Inc
Warning

012642-03 012642-06
AA12642-03EA 112 USD InStock
AA12642-03 AA12642-06
Potassium hexaiodoplatinate(IV) Pt ≥ 18,2%
Potassium hexaiodoplatinate(IV)
MDL: MFCD00049660

CAS No.: 16905-14-9 Molecular Formula: K2PtI6 Formula Weight: 1034.72 Physical Form: Crystalline MDL No.: MFCD00049660


Formula: I₆K₂Pt
Storage Temperature: Ambient
MDL Number: MFCD00049660
CAS Number: 16905-14-9
EINECS: 240-954-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty