Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper ≥99.9% (metals basis), plate, Thickness 6.35 mm (0.25 in)

Supplier: Origen Biomedical, Inc

013376-RR 013376-CU
AA13376-RREA 138 USD InStock
AA13376-RR AA13376-CU
Copper ≥99.9% (metals basis), plate, Thickness 6.35 mm (0.25 in)
Copper
MDL: MFCD00010965

Purity: 99.9% CAS No.: 7440-50-8 Physical Form: ≈569g/10x10cm Odor: Odorless MDL No.: MFCD00010965


Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty