Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium dichromate ≥99.0% ACS

Supplier: Origen Biomedical, Inc
Danger

013450-A4 013450-36
AA13450-A4EA 375 USD InStock
AA13450-A4 AA13450-22 AA13450-36
Potassium dichromate ≥99.0% ACS
Potassium dichromate
As an oxidizing agent, in dyeing, painting, decorating porcelain, lithography, in electric batteries, electroplating

Purity: 99.0% min CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Crystalline Density: 2.676 Appearance: Orange Odor: Odorless Melting Point: 398° Solubility: Soluble in water Merck Reference: 14,7627 UN No.: 3087 MDL No.: MFCD00011367

As an oxidizing agent, in dyeing, painting, decorating porcelain, lithography, in electric batteries, electroplating


Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Density: 2.676
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3087
ADR: 5.1 II
REACH: 01-2119454792-32
Merck Index: 14,7627
Order Now
Specification Test Results
Insoluble matter 0.005%
Loss on drying 0.05%
Cl 0.001%
SO3 0.005%
Ca 0.003%
Fe 0.001%
NA 0.02%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty