Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Zinc ≥99.9% (metals basis), soluble -325 mesh

Supplier: Origen Biomedical, Inc
Danger

013789-30 013789-A1
AA13789-30EA 72.69 USD InStock
AA13789-30 AA13789-A1
Zinc ≥99.9% (metals basis), soluble -325 mesh
Zinc
MDL: MFCD00011291

Purity: 99.9% CAS No.: 7440-66-6 Odor: Odorless UN No.: 1436 MDL No.: MFCD00011291


Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3 II
Merck Index: 12,10255
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty