Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Germanium(II) iodide ≥99.99% (metals basis)

Supplier: Origen Biomedical, Inc
Danger

014013-06 014013-03
AA14013-06EA 947 USD InStock
AA14013-06 AA14013-03
Germanium(II) iodide ≥99.99% (metals basis)
Germanium(II) iodide
MDL: MFCD00135537

Purity: 99.99% CAS No.: 13573-08-5 Molecular Formula: GeI2 Formula Weight: 326.40 Physical Form: -10 Mesh Density: 5.37 Appearance: Yellow to brown UN No.: 3260 MDL No.: MFCD00135537


Formula: GeI₂
Melting Pt: 460 °C
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135537
CAS Number: 13573-08-5
EINECS: 236-998-1
UN: 3260
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty