022921-AP
022921-K7
022921-M1
022921-K2
AA22921-APEA
64.17
USD
InStock
AA22921-AP
AA22921-K7
AA22921-M1
AA22921-K2
Chlorobenzene ≥99.9% for spectrophotometry
Chlorobenzene
MDL: MFCD00000530
Beilstein Registry No.: 605632
Purity: 99.9%
CAS No.: 108-90-7
Molecular Formula: C6H5Cl
Formula Weight: 112.56
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 1.106
Refractive Index: 1.5240
Appearance: Colorless
Odor: Aromatic
Melting Point: -45°
Boiling Point: 132°
Beilstein Reference: 605632
Merck Reference: 14,2121
UN No.: 1134
MDL No.: MFCD00000530