Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Molybdic acid ≥85% (MoO₃ basis) ACS

Supplier: Origen Biomedical, Inc
Warning

022983-36 022983-A3
AA22983-36EA 281.58 USD InStock
AA22983-36 AA22983-A3
Molybdic acid ≥85% (MoO₃ basis) ACS
Molybdic acid
MDL: MFCD00036287

CAS No.: 7782-91-4 Physical Form: Crystalline powder Density: 3.1 Odor: Odorless MDL No.: MFCD00036287


Formula: H₂MoO₄
MW: 161.95 g/mol
Boiling Pt: 300
Melting Pt: 300
Density: 3.1
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262
Order Now
Specification Test Results
Insoluble in dilute ammonium hydroxide 0.01%
Cl 0.002%
Arsenate phosphate and silicate (as SiO2) 0.001%
PO3 5 ppm
SO3 0.2%
Heavy Metals (as Pb) 0.003%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty