034108-06
034108-14
AA34108-06EA
446
USD
InStock
AA34108-06
AA34108-14
Dichlorodiethylgermane
Dichlorodiethylgermane
As an intermediate
CAS No.: 13314-52-8
Molecular Formula: (C2H5)2GeCl2
Formula Weight: 201.64
Flash Point: 63°(145°F)
Physical Form: Liquid
Density: 1.372
Refractive Index: 1.4710
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Acrid
Melting Point: -37°
Boiling Point: 175°
Solubility: Decomposes in water and NH3. Soluble in organic solvents
UN No.: 3265
MDL No.: MFCD00013589
As an intermediate