Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium oxide

Supplier: Origen Biomedical, Inc
Danger

036712-22 036712-14
AA36712-22EA 328.84 USD InStock
AA36712-22 AA36712-14
Disodium oxide
di-Sodium oxide
MDL: MFCD00046201

CAS No.: 1313-59-3 Molecular Formula: Na2O + Na2O2 Formula Weight: 61.98 Physical Form: Beads Density: 2.27 Sensitivity: Air and Moisture Sensitive Appearance: White to grey Odor: Odorless Melting Point: 400° dec. Merck Reference: 14,8651 UN No.: 1825 MDL No.: MFCD00046201


Formula: Na₂O
MW: 61.98 g/mol
Melting Pt: >400 °C (dec.)
Density: 2.27
Storage Temperature: Ambient
MDL Number: MFCD00046201
CAS Number: 1313-59-3
EINECS: 215-208-9
UN: 3085
ADR: 5.1 I
Merck Index: 14,8651
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty