Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Octadecane ≥90%, tech.

Supplier: Origen Biomedical, Inc
Danger

044301-A1 044301-A7
AA44301-A1EA 59.2 USD InStock
AA44301-A1 AA44301-A7
n-Octadecane ≥90%, tech.
n-Octadecane
MDL: MFCD00009007

Purity: 90% CAS No.: 593-45-3 Molecular Formula: CH3(CH2)16CH3 Formula Weight: 254.50 Flash Point: 166°(330°F) Physical Form: Crystalline mass/melt Density: 0.782 Refractive Index: 1.4390 Appearance: White Odor: Odorless Melting Point: 28° Boiling Point: 317° MDL No.: MFCD00009007


Formula: H₃C(CH₂)₁₆CH₃
MW: 254.5 g/mol
Boiling Pt: 316.1
Melting Pt: 28 °C
Density: 0.782
Flash Pt: 166 °C (330 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009007
CAS Number: 593-45-3
EINECS: 209-790-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty