071145-18
071145-09
AA71145-18EA
271
USD
InStock
AA71145-18
AA71145-09
Tetramethyltin 98%
Tetramethyltin
MDL: MFCD00008278
Insoluble in water. Soluble in organic solvents
Purity: 98%
CAS No.: 594-27-4
Molecular Formula: (CH3)4Sn
Formula Weight: 178.83
Flash Point: -12°(9°F)
Physical Form: Liquid
Density: 1.291
Refractive Index: 1.4410
Odor: Unpleasant
Melting Point: -54°
Boiling Point: 74-75°
Solubility: Insoluble in water. Soluble in organic solvents
UN No.: 3384
MDL No.: MFCD00008278