Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Titanium(III) oxide ≥99.8% (metals basis)

Supplier: Origen Biomedical, Inc

077127-30 077127-18
AA77127-30EA 935 USD InStock
AA77127-30 AA77127-18
Titanium(III) oxide ≥99.8% (metals basis)
Titanium(III) oxide
Purity: 99.8% CAS No.: 1344-54-3 Molecular Formula: Ti2O3 Formula Weight: 143.80 Physical Form: -325 Mesh Powder Density: 4.6 Melting Point: 2130° dec. MDL No.: MFCD00036278


Formula: O₃Ti₂
MW: 143.76
Melting Pt: 1900 (dec.)
Density: 4.6
Storage Temperature: Ambient
MDL Number: MFCD00036278
CAS Number: 1344-54-3
EINECS: 215-697-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty