Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lead(II) sulphide ≥99.9% (metals basis)

Supplier: Origen Biomedical, Inc
Danger

088284-18 088284-09
AA88284-18EA 415 USD InStock
AA88284-18 AA88284-09
Lead(II) sulphide ≥99.9% (metals basis)
Lead(II) sulphide
MDL: MFCD00016280

Purity: 99.9% CAS No.: 1314-87-0 Molecular Formula: PbS Formula Weight: 239.25 Physical Form: -200 Mesh Powder Density: 7.5 Refractive Index: 3.921 Odor: Odorless Melting Point: 1114° Merck Reference: 14,5421 UN No.: 3077 MDL No.: MFCD00016280


Formula: ̍PbS
MW: 239.27 g/mol
Boiling Pt: 1344
Melting Pt: 1114
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00016280
CAS Number: 1314-87-0
EINECS: 215-246-6
UN: 3077
ADR: 9 III
Merck Index: 14,5421
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty