Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophthalic acid 97%, Technical Grade

Supplier: Origen Biomedical, Inc
Warning

A10368-06 A10368-14 A10368-18
AAA10368-06EA 30.1 USD InStock
AAA10368-06 AAA10368-14 AAA10368-18
4-Nitrophthalic acid 97%, Technical Grade
4-Nitrophthalic acid
MDL: MFCD00007252 Beilstein Registry No.: 1882262


Formula: C₈H₅NO₆
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00007252
CAS Number: 610-27-5
EINECS: 210-215-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty