A10817-36
A10817-0E
A10817-22
AAA10817-36EA
195
USD
InStock
AAA10817-36
AAA10817-0E
AAA10817-22
Biphenyl-4-ol 98%
Biphenyl-4-ol
MDL: MFCD00002347
Beilstein Registry No.: 1907452
Purity: 98%
CAS No.: 92-69-3
Molecular Formula: C12H10O
Formula Weight: 170.21
Flash Point: 165°(329°F)
Melting Point: 164-168°
Boiling Point: 305-308°
Beilstein Reference: 1907452
Merck Reference: 14,7305
UN No.: 3077
MDL No.: MFCD00002347