Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Chloroacetanilide 98+%

Supplier: Origen Biomedical, Inc
Warning

A11214-22 A11214-14
AAA11214-22EA 146 USD InStock
AAA11214-22 AAA11214-36 AAAA11214-14
4'-Chloroacetanilide 98+%
4'-Chloroacetanilide
MDL: MFCD00000612 Beilstein Registry No.: 509638

Purity: 98+% CAS No.: 539-03-7 Molecular Formula: C8H8ClNO Formula Weight: 169.61 Melting Point: 178-180° Boiling Point: 333° Merck Reference: 14,2111 MDL No.: MFCD00000612


Formula: C₈H₈ClNO
MW: 169.61 g/mol
Boiling Pt: 333 °C
Melting Pt: 178…180 °C
Density: 1.385
Storage Temperature: Ambient
MDL Number: MFCD00000612
CAS Number: 539-03-7
EINECS: 208-707-8
Merck Index: 14,2111
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty