A11453-14
A11453-06
A11453-22
AAA11453-14EA
128
USD
InStock
AAA11453-14
AAA11453-06
AAA11453-22
Perfluorotoluene 98+%
Perfluorotoluene
MDL: MFCD00000375
Beilstein Registry No.: 2057072
Fieser: 21,338
Purity: 98+%
CAS No.: 434-64-0
Molecular Formula: C7F8
Formula Weight: 236.06
Flash Point: 20°(68°F)
Refractive Index: 1.3680
Melting Point: -66°
Boiling Point: 103-105°
Beilstein Reference: 2057072
UN No.: 1992
MDL No.: MFCD00000375