A11554-0B
A11554-30
AAA11554-0BEA
226
USD
InStock
AAA11554-0B
AAA11554-30
Dibenzylamine 98%
Dibenzylamine
MDL: MFCD00004770
Beilstein Registry No.: 909664
Purity: 98%
CAS No.: 103-49-1
Molecular Formula: C14H15N
Formula Weight: 197.28
Flash Point: 143°(289°F)
Refractive Index: 1.5745
Odor: Weak
pH: 8.9
Melting Point: -26°
Boiling Point: 300°
Beilstein Reference: 909664
Merck Reference: 14,3011
UN No.: 2735
MDL No.: MFCD00004770