Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Chloropropiophenone 98%

Supplier: Origen Biomedical, Inc
Warning

A11633-14 A11633-22
AAA11633-14EA 46.9 USD InStock
AAA11633-14 AAA11633-22
4'-Chloropropiophenone 98%
4'-Chloropropiophenone
MDL: MFCD00000626 Beilstein Registry No.: 1100638 Insoluble in water

Purity: 98% CAS No.: 6285-05-8 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 33-36° Boiling Point: 125-126°/13mm Solubility: Insoluble in water Beilstein Reference: 1100638 MDL No.: MFCD00000626


Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 277
Melting Pt: 33…36 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000626
CAS Number: 6285-05-8
EINECS: 228-511-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty