A13281-18
A13281-30
AAA13281-18EA
39
USD
InStock
AAA13281-18
AAA13281-30
2-Benzoylbenzoic acid 98+%
2-Benzoylbenzoic acid
Purity: 98+%
CAS No.: 85-52-9
Molecular Formula: C14H10O3
Formula Weight: 226.23
Odor: Odorless
Melting Point: 127-130°
Boiling Point: 257°
Beilstein Reference: 1107841
MDL No.: MFCD00002472