Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Uric acid 99%

Supplier: Origen Biomedical, Inc

A13346-22 A13346-36 A13346-14
AAA13346-22EA 134.7 USD InStock
AAA13346-22 AAA13346-36 AAA13346-14
Uric acid 99%
Uric acid
Purity: 99% CAS No.: 69-93-2 Molecular Formula: C5H4N4O3 Formula Weight: 168.11 Melting Point: 300° Beilstein Reference: 156158 Merck Reference: 14,9875 MDL No.: MFCD00005712


Formula: C₅H₄N₄O₃
MW: 168.11 g/mol
Melting Pt: 436
Density: 1.89
Storage Temperature: Ambient
MDL Number: MFCD00005712
CAS Number: 69-93-2
EINECS: 200-720-7
Merck Index: 14,9875
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty