A13367-22
A13367-06
AAAA13367-22EA
985
USD
InStock
AAAA13367-22
AAA13367-06
4-Hexyloxybenzoyl chloride 98%
4-Hexyloxybenzoyl chloride
Purity: 98%
CAS No.: 39649-71-3
Molecular Formula: C13H17ClO2
Formula Weight: 240.73
Flash Point: 110°(230°F)
Refractive Index: 1.5383
Boiling Point: 203-205°/15mm
Beilstein Reference: 2210619
UN No.: 3265
MDL No.: MFCD00000690