A13575-06
A13575-14
A13575-22
AAA13575-06EA
30.2
USD
InStock
AAA13575-06
AAA13575-14
AAA13575-22
Pentafluorobenzonitrile 98%
Pentafluorobenzonitrile
Purity: 98%
CAS No.: 773-82-0
Molecular Formula: C7F5N
Formula Weight: 193.07
Flash Point: 29°(84°F)
Refractive Index: 1.4425
Melting Point: 1-2°
Boiling Point: 161-162°
Beilstein Reference: 1913453
UN No.: 1992
MDL No.: MFCD00001775