Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorotriphenylsilane 96%

Supplier: Origen Biomedical, Inc
Danger

A13678-18 A13678-22 A13678-14 A13678-06
AAAA13678-18EA 156 USD InStock
AAAA13678-18 AAA13678-22 AAA13678-14 AAA13678-06
Chlorotriphenylsilane 96%
Chlorotriphenylsilane
Purity: 96% CAS No.: 76-86-8 Molecular Formula: C18H15ClSi Formula Weight: 294.86 Flash Point: 200°(392°F) Odor: Pungent Boiling Point: 188-189°/3mm Beilstein Reference: 1820487 UN No.: 3261 MDL No.: MFCD00000496


Formula: C₁₈H₁₅ClSi
MW: 294.86 g/mol
Boiling Pt: 188…189 °C (3 mmHg)
Melting Pt: 91…96 °C
Flash Pt: >200 °C (392 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000496
CAS Number: 76-86-8
EINECS: 200-989-0
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty