A14692-14
A14692-22
A14692-36
AAA14692-14EA
23.49
USD
InStock
AAA14692-14
AAA14692-22
AAA14692-36
Isobutyl chloroformate 98%
Isobutyl chloroformate
Purity: 98%
CAS No.: 543-27-1
Molecular Formula: C5H9ClO2
Formula Weight: 136.58
Flash Point: 30°(86°F)
Refractive Index: 1.4060
Melting Point: -80°
Boiling Point: 128-129°
Beilstein Reference: 956590
Merck Reference: 14,5139
UN No.: 2742
MDL No.: MFCD00000642