Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(-)-Leucine 99%

Supplier: Origen Biomedical, Inc

A14842-06 A14842-14
AAA14842-06EA 64.7 USD InStock
AAA14842-06 AAA14842-14 AAAA14842-22
D-(-)-Leucine 99%
D-(-)-Leucine
Purity: 99% CAS No.: 328-38-1 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721721


Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00063088
CAS Number: 328-38-1
EINECS: 206-327-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty