Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromophenol 98%

Supplier: Origen Biomedical, Inc
Warning

A14849-09 A14849-18 A14849-30
AAA14849-09EA 52.78 USD InStock
AAA14849-09 AAA14849-18 AAA14849-30
3-Bromophenol 98%
3-Bromophenol
Purity: 98% CAS No.: 591-20-8 Molecular Formula: C6H5BrO Formula Weight: 173.01 Flash Point: 110°(230°F) Refractive Index: 1.5972 Melting Point: 28-32° Boiling Point: 235-236° Beilstein Reference: 1853950 Merck Reference: 14,1428 MDL No.: MFCD00002253


Formula: C₆H₅BrO
Boiling Pt: 235…236 °C
Melting Pt: 28…32 °C
Density: 0.641 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002253
CAS Number: 591-20-8
EINECS: 209-706-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,01413
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty