A15067-0F
A15067-AE
A15067-AP
AAA-A15067-0FEA
960
USD
InStock
AAA-A15067-0F
AA-A15067-AE
AA-A15067-AP
Trioctylamine 95%
Trioctylamine
Purity: 95%
CAS No.: 1116-76-3
Molecular Formula: C24H51N
Formula Weight: 353.68
Storage Temperature: 2°C to 8°C
Flash Point: 158°(316°F)
Refractive Index: 1.4485
Odor: Amine-like
Melting Point: -39°
Boiling Point: 164-168°/0.7mm
Beilstein Reference: 1210618
UN No.: 3082
MDL No.: MFCD00009560