Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxyphenyl isothiocyanate 98%

Supplier: Origen Biomedical, Inc
Danger

A15090-06 A15090-22 A15090-14
AAA15090-06EA 46.1 USD InStock
AAA15090-06 AAA15090-22 AAA15090-14
3-Methoxyphenyl isothiocyanate 98%
3-Methoxyphenyl isothiocyanate
Purity: 98% CAS No.: 3125-64-2 Molecular Formula: C8H7NOS Formula Weight: 165.22 Flash Point: 110°(230°F) Refractive Index: 1.6390 Odor: Pungent Boiling Point: 133-134°/10mm Beilstein Reference: 775987 UN No.: 2922 MDL No.: MFCD00040335


Formula: C₈H₇NOS
Storage Temperature: Ambient
MDL Number: MFCD00040335
CAS Number: 3125-64-2
UN: 2922
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty