Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,8-Dibromooctane 98%

Supplier: Origen Biomedical, Inc

A15651-14 A15651-22 A15651-18
AAAA15651-14EA 65.7 USD InStock
AAAA15651-14 AAAA15651-22 AAA15651-18
1,8-Dibromooctane 98%
1,8-Dibromooctane
Purity: 98% CAS No.: 4549-32-0 Molecular Formula: Br(CH2)8Br Formula Weight: 272.02 Flash Point: 110°(230°F) Refractive Index: 1.4995 Melting Point: 15-16° Boiling Point: 270-272° Beilstein Reference: 1698114 MDL No.: MFCD00000277


Formula: BrH₂C(CH₂)₆CH₂Br
Boiling Pt: 270…272 °C
Melting Pt: 15…16 °C
Density: 1.46 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000277
CAS Number: 4549-32-0
EINECS: 224-912-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty