Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetamidobenzoic acid 98%

Supplier: Origen Biomedical, Inc

A15902-14 A15902-22
AAA15902-14EA 29.2 USD InStock
AAA15902-14 AAA15902-22 AAA15902-36
3-Acetamidobenzoic acid 98%
3-Acetamidobenzoic acid
Purity: 98% CAS No.: 587-48-4 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 249-252° Beilstein Reference: 2210867


Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 249…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00013983
CAS Number: 587-48-4
EINECS: 209-600-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty