Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Acetylindole 98%

Supplier: Origen Biomedical, Inc

A16221-14 A16221-06 A16221-22
AAA16221-14EA 210 USD InStock
AAA16221-14 AAA16221-06 AAA16221-22
3-Acetylindole 98%
3-Acetylindole
Purity: 98% CAS No.: 703-80-0 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 186-192° Beilstein Reference: 122579


Formula: C₁₀H₉NO
Boiling Pt: 144 - 145 °C (10 -rr)
Melting Pt: 186…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00005626
CAS Number: 703-80-0
EINECS: 211-875-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty