A17991-06
A17991-09
A17991-18
AAAA17991-06EA
29.2
USD
InStock
AAAA17991-06
AAA17991-09
AAA17991-18
2,2-Diphenylpropiononitrile 97%
2,2-Diphenylpropiononitrile
Purity: 97%
CAS No.: 5558-67-8
Molecular Formula: C15H13N
Formula Weight: 207.28
Flash Point: 110°(230°F)
Refractive Index: 1.5720
Boiling Point: 144-145°/2mm
Beilstein Reference: 1955952
UN No.: 3276
MDL No.: MFCD00001846