Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-bromohexanoate 97+%

Supplier: Origen Biomedical, Inc
Warning

A18398-09 A18398-30 A18398-18
AAA18398-09EA 38 USD InStock
AAA18398-09 AAA18398-30 AAA18398-18
Ethyl-6-bromohexanoate 97+%
Ethyl-6-bromohexanoate
Purity: 97+% CAS No.: 25542-62-5 Molecular Formula: Br(CH2)5CO2CH2CH3 Formula Weight: 223.12 Flash Point: 110°(230°F) Refractive Index: 1.4595 Boiling Point: 128-130°/16mm Beilstein Reference: 1756051 MDL No.: MFCD00000270


Formula: BrH₂C(CH₂)₄COOCH₂CH₃
Boiling Pt: 69 - 72 °C (0.1 -rr)
Melting Pt: 33 °C
Density: 1.2592 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000270
CAS Number: 25542-62-5
EINECS: 247-085-2
UN: 3272
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty