A18952-36
A18952-22
A18952-14
AAA18952-36EA
215.92
USD
InStock
AAA18952-36
AAA18952-22
AAAA18952-14
2-Aminothiophenol 98%
2-Aminothiophenol
Purity: 98%
CAS No.: 137-07-5
Molecular Formula: C6H7NS
Formula Weight: 125.19
Flash Point: 79°(174°F)
Refractive Index: 1.6420
Odor: Stench
Melting Point: 18-23°
Boiling Point: 234°
Beilstein Reference: 606076
UN No.: 3267
MDL No.: MFCD00007702