Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Nitrophenyl)acetonitrile 98%

Supplier: Origen Biomedical, Inc
Warning

B20190-06 B20190-14
AAB20190-06EA 81 USD InStock
AAB20190-06 AAB20190-14 AAB20190-03
(2-Nitrophenyl)acetonitrile 98%
(2-Nitrophenyl)acetonitrile
Purity: 98% CAS No.: 610-66-2 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 81-85° Boiling Point: 178°/12mm UN No.: 3439 MDL No.: MFCD00007183


Formula: C₈H₆N₂O₂
MW: 162.15
Boiling Pt: 178 °C/12 mm
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007183
CAS Number: 610-66-2
EINECS: 210-231-0
UN: 3439
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty