B21340-30
B21340-0B
AAB21340-30EA
48
USD
InStock
AAB21340-30
AAB21340-0B
Benzotrifluoride 99%
Benzotrifluoride
Purity: 99%
CAS No.: 98-08-8
Molecular Formula: C7H5F3
Formula Weight: 146.11
Flash Point: 12°(54°F)
Refractive Index: 1.4145
Odor: Aromatic
Melting Point: -29°
Boiling Point: 101-103°
Beilstein Reference: 1906908
Merck Reference: 14,1110
UN No.: 2338
MDL No.: MFCD00000372