Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butoxybenzoic acid 98%

Supplier: Origen Biomedical, Inc

B21506-22 B21506-14
AAB21506-22EA 192 USD InStock
AAB21506-22 AAB21506-14
4-Butoxybenzoic acid 98%
4-Butoxybenzoic acid
Purity: 98% CAS No.: 1498-96-0 Molecular Formula: C11H14O3 Formula Weight: 194.23 Melting Point: 146-147° (C-N) MDL No.: MFCD00002546


Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 319
Melting Pt: 149 (C-Nematic)
Storage Temperature: Ambient
MDL Number: MFCD00002546
CAS Number: 1498-96-0
EINECS: 216-104-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty