B21981-06
B21981-03
B21981-14
AAB21981-06EA
151.96
USD
InStock
AAB21981-06
AAB21981-03
AAB21981-14
4-Allyl-2,6-dimethoxyphenol 98%
4-Allyl-2,6-dimethoxyphenol
Purity: 98%
CAS No.: 6627-88-9
Molecular Formula: C11H14O3
Formula Weight: 194.23
Flash Point: 110°(230°F)
Refractive Index: 1.5480
Boiling Point: 168-169°/11mm
MDL No.: MFCD00008655