Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Methylenanthic acid 98%

Supplier: Origen Biomedical, Inc
Danger

B22050-14 B22050-22 B22050-06
AAB22050-14EA 77 USD InStock
AAB22050-14 AAB22050-22 AAB22050-06
(±)-2-Methylenanthic acid 98%
(±)-2-Methylenanthic acid
Purity: 98% CAS No.: 1188-02-9 Molecular Formula: C8H16O2 Formula Weight: 144.22 Flash Point: 110°(230°F) Refractive Index: 1.4255 Boiling Point: 139-141°/30mm MDL No.: MFCD00142944


Formula: C₈H₁₆O₂
Boiling Pt: 139…141 °C/30 mm
Density: 0.9122 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142944
CAS Number: 1188-02-9
EINECS: 214-704-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty