Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.1%
B22353-22
B22353-06
B22353-14
AAB22353-22EA
972
USD
InStock
AAB22353-22
AAB22353-06
AAB22353-14
4-Methoxystyrene 98% stabilized
4-Methoxystyrene
Purity: 98%
CAS No.: 637-69-4
Molecular Formula: C9H10O
Formula Weight: 134.18
Storage Temperature: 2°C to 8°C
Flash Point: 72°(161°F)
Refractive Index: 1.5610
Boiling Point: 204-205°
Beilstein Reference: 1098935
MDL No.: MFCD00008619