4-(Trifluoromethyl)pyridine 97%
Supplier: Origen Biomedical, Inc
|
Danger
|
Purity: 97%
CAS No.: 3796-24-5
Molecular Formula: C6H4F3N
Formula Weight: 147.10
Flash Point: 20°(68°F)
Refractive Index: 1.4147
Boiling Point: 107-110°
Beilstein Reference: 1364589
UN No.: 1993
MDL No.: MFCD00153289
Formula:
C₆H₄F₃N MW: 147.1 Boiling Pt: 107…110 °C Density: 1.270 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number: |