Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromophenol 98%

Supplier: Origen Biomedical, Inc
Danger

B23394-22 B23394-14
AAAB23394-36EA 543.9 USD InStock
AAAB23394-36 AAB23394-22 AAB23394-14
2,4-Dibromophenol 98%
2,4-Dibromophenol
Purity: 98% CAS No.: 615-58-7 Molecular Formula: C6H4Br2O Formula Weight: 251.92 Flash Point: 110°(230°F) Odor: Phenol-like Boiling Point: 239° Beilstein Reference: 1861291 UN No.: 2811 MDL No.: MFCD00002149


Formula: C₆H₄Br₂O
Boiling Pt: 239 °C
Melting Pt: 35…38 °C
Density: 2.07 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002149
CAS Number: 615-58-7
EINECS: 210-436-5
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty