B23709-06
B23709-03
B23709-14
AAB23709-06EA
133
USD
InStock
AAB23709-06
AAB23709-03
AAB23709-14
4-Vinylphenylboronic acid 97%
4-Vinylphenylboronic acid
Purity: 97%
CAS No.: 2156-04-9
Molecular Formula: C8H9BO2
Formula Weight: 147.97
Storage Temperature: 2°C to 8°C
Melting Point: ca 190°
Beilstein Reference: 2829387
MDL No.: MFCD00239441