Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl isophthalate 98%

Supplier: Origen Biomedical, Inc

B23779-22 B23779-36 B23779-0E
AAB23779-22EA 18.4 USD InStock
AAB23779-22 AAB23779-36 AAB23779-0E
Dimethyl isophthalate 98%
Dimethyl isophthalate
Purity: 98% CAS No.: 1459-93-4 Molecular Formula: C10H10O4 Formula Weight: 194.19 Flash Point: 138°(280°F) Odor: Mild Boiling Point: 282° Beilstein Reference: 1912251 MDL No.: MFCD00008433


Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 285
Melting Pt: 67…70 °C
Density: 1.194
Flash Pt: 138 °C(280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008433
CAS Number: 1459-93-4
EINECS: 215-951-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty