Special Application Instrument Thermometers, Thermco®
Supplier: Thermco
ACC603FC ACC6113SFC ACC715S ACC715SSC ACC6471S ACC647SSC ACC717S ACC717SSC ACC6371S ACC6371SSC
89137-090EA 462.53 USD InStock
89137-090 89137-092 89137-094 89137-096 89137-098 89137-100 89137-102 89137-104 89137-106 89137-108
Special Application Instrument Thermometers, Thermco®
Thermometers Analog Thermometers
These non-mercury thermometers are calibrated against NIST standards to assure accuracy and reproducibility
Some thermometers have fluoropolymer coating. The certified thermometers include a deluxe, felt-lined storage box. Thermometer styles available include General-Purpose Certified Thermometers; Thermometers for Block Heaters; Thermometers for Blood Bank Refrigerators; and Thermometers for Water baths and Incubators.
Order Now
ORDER
SPECIFICATIONS
SERVICIOS
Servicios: VWRCATALYST™ offers recertification, calibration, and repair services for all makes and models of thermometers. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for quote today.
Our thermometer services include:
Complete visual inspection for any physical damage. If possible, the damage will be repaired.
Recertification from 1 Point to 15 Points.
Thermometer readings are compared against NIST standards in high precision calibration baths, using the comparison method at determined test temperatures.
Each temperature is recorded to 1/10th of a scale division.
A calibration report is generated, detailing pertinent information including the correction factors for each test point.
All calibration information is entered into a data storage system.
All certification procedures are performed in accordance with NIST and ASTM guidelines.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty